missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetone, 97%, Thermo Scientific Chemicals

Catalog Number p-3212031
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
6186-22-7
C24H24N2O5S
452.53
MFCD00210401
CDUPPXGKUKTGLU-UHFFFAOYSA-N
4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone
736332
COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1
This item is not returnable. View return policy
6186-22-7
C24H24N2O5S
452.53
MFCD00210401
CDUPPXGKUKTGLU-UHFFFAOYSA-N
4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone
736332
COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5560 to 1.5590 (20°C, 589nm)
Quantity 5 g
IUPAC Name 1-(4-bromophenyl)propan-2-one
Formula Weight 213.08
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 4-Bromophenylacetone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'