missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl methyl sulfone, 99%, Thermo Scientific™

Catalog Number 10561282
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3466-32-8
C7H7BrO2S
235.1
MFCD00025065
FJLFSYRGFJDJMQ-UHFFFAOYSA-N
4-bromophenyl methyl sulfone, 1-bromo-4-methylsulfonyl benzene, 1-bromo-4-methanesulfonylbenzene, p-bromophenyl methyl sulfone, methyl 4-bromophenyl sulfone, 4-bromophenyl methyl sulphone, benzene, 1-bromo-4-methylsulfonyl, 4-bromophenylmethylsulfone, p-methyl benzenesulfobromide, sulfone, p-bromophenyl methyl
77014
1-bromo-4-methylsulfonylbenzene
CS(=O)(=O)C1=CC=C(C=C1)Br
This item is not returnable. View return policy
3466-32-8
C7H7BrO2S
235.1
MFCD00025065
FJLFSYRGFJDJMQ-UHFFFAOYSA-N
4-bromophenyl methyl sulfone, 1-bromo-4-methylsulfonyl benzene, 1-bromo-4-methanesulfonylbenzene, p-bromophenyl methyl sulfone, methyl 4-bromophenyl sulfone, 4-bromophenyl methyl sulphone, benzene, 1-bromo-4-methylsulfonyl, 4-bromophenylmethylsulfone, p-methyl benzenesulfobromide, sulfone, p-bromophenyl methyl
77014
1-bromo-4-methylsulfonylbenzene
CS(=O)(=O)C1=CC=C(C=C1)Br
CAS Min % 98.5
CAS Max % 100.0
Melting Point 102°C to 105°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 235.1
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromophenyl methyl sulfone
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromophenyl methyl sulfone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'