missing translation for 'onlineSavingsMsg'
Learn More

4-bromophenyl isothiocyanate, Thermo Scientific™

Catalog Number p-4391228
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
1985-12-2
C7H4BrNS
214.08
XQACWEBGSZBLRG-UHFFFAOYSA-N
4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
16133
1-bromo-4-isothiocyanatobenzene
C1=CC(=CC=C1N=C=S)Br
This item is not returnable. View return policy
1985-12-2
C7H4BrNS
214.08
XQACWEBGSZBLRG-UHFFFAOYSA-N
4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
16133
1-bromo-4-isothiocyanatobenzene
C1=CC(=CC=C1N=C=S)Br
Assay Percent Range 97%
Quantity 10 g
Formula Weight 214.09
Percent Purity 97%
Chemical Name or Material 4-Bromophenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'