missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenyl)cyclopropylmethanone, 95%, Thermo Scientific Chemicals

Catalog Number p-3777708
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6952-89-2
MFCD00019227
QTHHOINSCNBYQO-UHFFFAOYSA-N
4-bromophenyl cyclopropyl methanone, 4-bromophenyl cyclopropyl ketone, 4-bromophenyl cyclopropylmethanone, methanone, 4-bromophenyl cyclopropyl, 4-bromo-cyclopropylcarbonylbenzene, acmc-1bag8, 4-bromobenzoyl cyclopropane, bromophenylcyclopropylmethanone
81394
(4-bromophenyl)-cyclopropylmethanone
C1CC1C(=O)C2=CC=C(C=C2)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6952-89-2
MFCD00019227
QTHHOINSCNBYQO-UHFFFAOYSA-N
4-bromophenyl cyclopropyl methanone, 4-bromophenyl cyclopropyl ketone, 4-bromophenyl cyclopropylmethanone, methanone, 4-bromophenyl cyclopropyl, 4-bromo-cyclopropylcarbonylbenzene, acmc-1bag8, 4-bromobenzoyl cyclopropane, bromophenylcyclopropylmethanone
81394
(4-bromophenyl)-cyclopropylmethanone
C1CC1C(=O)C2=CC=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Beige to Orange or Yellow
Density 1.4500g/mL
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C10H9BrO
Refractive Index 1.5919
Quantity 50 g
Specific Gravity 1.45
Molecular Weight (g/mol) 225.09
Formula Weight 225.09
Percent Purity 95%
Physical Form Crystalline Powder or Crystalline Low Melting Mass
Chemical Name or Material (4-Bromophenyl)cyclopropylmethanone
Show More Show Less
compliance-icons
Product Identifier
  • (4-Bromophenyl)cyclopropylmethanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'