missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl acetate, 99%, Thermo Scientific™

Catalog Number p-4813082
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1927-95-3
C8H7BrO2
215.05
XEXHCQJRVMVJMY-UHFFFAOYSA-N
4-bromophenol acetate, p-bromophenyl acetate, phenol, 4-bromo-, acetate, p-acetoxybromobenzene, 4-bromoacetoxybenzene, phenol, 4-bromo-, 1-acetate, 4-bromophenyl acetate, phenol, p-bromo-, acetate, phenol, 4-bromo-,1-acetate, acetic acid 4-bromophenyl ester
74726
(4-bromophenyl) acetate
CC(=O)OC1=CC=C(C=C1)Br
This item is not returnable. View return policy
1927-95-3
C8H7BrO2
215.05
XEXHCQJRVMVJMY-UHFFFAOYSA-N
4-bromophenol acetate, p-bromophenyl acetate, phenol, 4-bromo-, acetate, p-acetoxybromobenzene, 4-bromoacetoxybenzene, phenol, 4-bromo-, 1-acetate, 4-bromophenyl acetate, phenol, p-bromo-, acetate, phenol, 4-bromo-,1-acetate, acetic acid 4-bromophenyl ester
74726
(4-bromophenyl) acetate
CC(=O)OC1=CC=C(C=C1)Br

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Assay Percent Range 98.5% min. (GC)
Quantity 5 g
Formula Weight 215.05
Percent Purity 99%
Chemical Name or Material 4-Bromophenyl acetate
compliance-icons compliance-icons
Product Identifier
  • 4-Bromophenyl acetate, 99%
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'