missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ (4-Bromophenoxy)-tert-butyldimethylsilane, 97%
Click to view available options
Quantity:
5 mL
25 mL
Specifications
Boiling Point | 137°C (25mmHg) |
Color | Colorless |
Physical Form | Liquid |
Quantity | 5 mL |
UN Number | UN1993 |
Formula Weight | 287.27 |
Percent Purity | 97% |
Refractive Index | 1.5110 |
Density | 1.174g/mL |
Chemical Name or Material | (4-Bromophenoxy)-tert-butyldimethylsilane |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'