missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromophenethyl bromide, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 1.7300g/mL |
Boiling Point | 97°C to 98.5°C (1.0 mmHg) |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (GC) |
Quantity | 10 g |
Specific Gravity | 1.73 |
Show More |
Safety and Handling
Product Identifier
- 4-Bromophenethyl bromide, 96%
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'