missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenethyl bromide, 96%, Thermo Scientific Chemicals

Catalog Number p-4258814
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
1746-28-7
C8H8Br2
263.96
APTDRDYSJZQPPI-UHFFFAOYSA-N
1-bromo-4-2-bromoethyl benzene, 4-bromophenethyl bromide, 4-bromophenethylbromide, benzene,1-bromo-4-2-bromoethyl, 4-bromo-1-2-bromoethyl benzene, pubchem16698, 4-bromobenzylmethyl bromide, acmc-209e8m, 1-bromo-4-2-bromoethyl ;benzene
11010825
1-bromo-4-(2-bromoethyl)benzene
C1=CC(=CC=C1CCBr)Br
This item is not returnable. View return policy
1746-28-7
C8H8Br2
263.96
APTDRDYSJZQPPI-UHFFFAOYSA-N
1-bromo-4-2-bromoethyl benzene, 4-bromophenethyl bromide, 4-bromophenethylbromide, benzene,1-bromo-4-2-bromoethyl, 4-bromo-1-2-bromoethyl benzene, pubchem16698, 4-bromobenzylmethyl bromide, acmc-209e8m, 1-bromo-4-2-bromoethyl ;benzene
11010825
1-bromo-4-(2-bromoethyl)benzene
C1=CC(=CC=C1CCBr)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.7300g/mL
Boiling Point 97°C to 98.5°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.73
Formula Weight 263.96
Percent Purity 96%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Bromophenethyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Bromophenethyl bromide, 96%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'