missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromonaphthalene-1-boronic acid, 97%

Brand:  Alfa Aesar™ H55625.03

Code : NEW

Additional Details : CAS Number : 145965-14-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15451938

  • £51.80 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromonaphthalene-1-boronic acid
145965-14-6
250.88
MFCD08701764

Specifications

4-Bromonaphthalene-1-boronic acid
230° to 260°C
Solid
97%
MFCD08701764
145965-14-6
White to pale brown
1g
250.88
Videos
SDS