missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)biphenyl, 96%, Thermo Scientific Chemicals

Catalog Number p-4510686
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2567-29-5
C13H11Br
247.13
HZQLUIZFUXNFHK-UHFFFAOYSA-N
4-bromomethylbiphenyl, 4-bromomethyl biphenyl, 4-bromomethyl-1,1'-biphenyl, 4-phenylbenzyl bromide, 1-bromomethyl-4-phenylbenzene, bromoethylbiphenyl, 4-biphenylmethylbromide, 4-bromomethyl-biphenyl
257716
1-(bromomethyl)-4-phenylbenzene
C1=CC=C(C=C1)C2=CC=C(C=C2)CBr
This item is not returnable. View return policy
2567-29-5
C13H11Br
247.13
HZQLUIZFUXNFHK-UHFFFAOYSA-N
4-bromomethylbiphenyl, 4-bromomethyl biphenyl, 4-bromomethyl-1,1'-biphenyl, 4-phenylbenzyl bromide, 1-bromomethyl-4-phenylbenzene, bromoethylbiphenyl, 4-biphenylmethylbromide, 4-bromomethyl-biphenyl
257716
1-(bromomethyl)-4-phenylbenzene
C1=CC=C(C=C1)C2=CC=C(C=C2)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 83.0°C to 86.0°C
Color Beige to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H4CH2Br
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene
Formula Weight 247.13
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-(Bromomethyl)biphenyl
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Bromomethyl)biphenyl
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'