missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Bromomethyl)-3-methyl-5-phenylisoxazole, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 4-bromomethyl-3-methyl-5-phenylisoxazole, 4-bromomethyl-3-methyl-5-phenyl-1,2-oxazole, pubchem23336, acmc-1c8sr, 4-bromomethyl-3-methyl-5-phenyl-isoxazole, isoxazole,4-bromomethyl-3-methyl-5-phenyl, isoxazole, 4-bromomethyl-3-methyl-5-phenyl |
Formula Weight | 252.11 |
Percent Purity | 95% |
Chemical Name or Material | 4-(Bromomethyl)-3-methyl-5-phenylisoxazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'