missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole 97%, Maybridge

Brand:  Maybridge CC64808DA

Code : 31

Additional Details : CAS Number : 912569-72-3 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10796915

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
96.5
97%
C12H10BrF3N2O
MFCD09879947
912569-72-3
100.0
97%
335.12

Specifications

4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
96.5
Amber Glass Bottle
97%
C12H10BrF3N2O
MFCD09879947
912569-72-3
100.0
1g
97%
335.12
Videos
SDS