missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisoquinoline, 98%, Thermo Scientific Chemicals

Catalog Number 10487090
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1532-97-4
C9H6BrN
208.06
MFCD00006904
SCRBSGZBTHKAHU-UHFFFAOYSA-N
isoquinoline, 4-bromo, 4-bromo-isoquinoline, 4-isoquinolinyl bromide, 4-bromisochinolin, 4bromoisoquinoline, 4-bromoisoquinoline, 4-bromo isoquinoline, 4-bromoisoquinoline?, pubchem6212, acmc-1bu1h
73743
4-bromoisoquinoline
BrC1=C2C=CC=CC2=CN=C1
This item is not returnable. View return policy
1532-97-4
C9H6BrN
208.06
MFCD00006904
SCRBSGZBTHKAHU-UHFFFAOYSA-N
isoquinoline, 4-bromo, 4-bromo-isoquinoline, 4-isoquinolinyl bromide, 4-bromisochinolin, 4bromoisoquinoline, 4-bromoisoquinoline, 4-bromo isoquinoline, 4-bromoisoquinoline?, pubchem6212, acmc-1bu1h
73743
4-bromoisoquinoline
BrC1=C2C=CC=CC2=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 39.0°C to 43.0°C
Color White to Beige
Boiling Point 280.0°C to 285.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 20, 384
Formula Weight 208.06
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromoisoquinoline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromoisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'