missing translation for 'onlineSavingsMsg'
Learn More

4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-74747
11.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
460-00-4
C6H4BrF
175.00
MFCD00000342
AITNMTXHTIIIBB-UHFFFAOYSA-N
4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
9993
1-bromo-4-fluorobenzene
FC1=CC=C(Br)C=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
460-00-4
C6H4BrF
175.00
MFCD00000342
AITNMTXHTIIIBB-UHFFFAOYSA-N
4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
9993
1-bromo-4-fluorobenzene
FC1=CC=C(Br)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -8.0°C
Color Colorless
Density 1.5930g/mL
Boiling Point 150.0°C to 153.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4F
Refractive Index 1.5260 to 1.5280
Quantity 10 mL
Beilstein 05, 209
Specific Gravity 1.593
Solubility Information Solubility in water: 0.14g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,acetone,soluble in ether
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromofluorobenzene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'