missing translation for 'onlineSavingsMsg'
Learn More

4-Bromodiphenyl ether, 99%, Thermo Scientific Chemicals

Catalog Number p-7043588
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
101-55-3
C12H9BrO
249.107
MFCD00000094
JDUYPUMQALQRCN-UHFFFAOYSA-N
4-bromodiphenyl ether, 4-bromophenyl phenyl ether, 4-bromophenoxybenzene, p-bromodiphenyl ether, p-phenoxybromobenzene, benzene, 1-bromo-4-phenoxy, p-bromophenyl phenyl ether, 4-bromodiphenylether, p-bromophenoxybenzene, ether, p-bromophenyl phenyl
7565
CHEBI:77421
1-bromo-4-phenoxybenzene
C1=CC=C(C=C1)OC2=CC=C(C=C2)Br
This item is not returnable. View return policy
101-55-3
C12H9BrO
249.107
MFCD00000094
JDUYPUMQALQRCN-UHFFFAOYSA-N
4-bromodiphenyl ether, 4-bromophenyl phenyl ether, 4-bromophenoxybenzene, p-bromodiphenyl ether, p-phenoxybromobenzene, benzene, 1-bromo-4-phenoxy, p-bromophenyl phenyl ether, 4-bromodiphenylether, p-bromophenoxybenzene, ether, p-bromophenyl phenyl
7565
CHEBI:77421
1-bromo-4-phenoxybenzene
C1=CC=C(C=C1)OC2=CC=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 17°C to 18°C
Density 1.42
Boiling Point 304°C to 305°C
Flash Point >110°C (230°F)
Refractive Index 1.608
Quantity 25 g
UN Number UN3082
Beilstein 1308972
Formula Weight 249.11
Percent Purity 99%
Chemical Name or Material 4-Bromodiphenyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'