missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzoyl chloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
4-Bromobenzoyl chloride is used in the synthesis of HIV-1 protease inhibitors. It is also used in the synthesis of oxazoles, cyclohexanone derivatives as analgesic and antimicrobial agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromobenzoyl chloride is used in the synthesis of HIV-1 protease inhibitors. It is also used in the synthesis of oxazoles, cyclohexanone derivatives as analgesic and antimicrobial agents.
Solubility
Soluble in methanol. Reacts with water forming Hcl.
Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases. Store at 2 - 8°C.
4-Bromobenzoyl chloride is used in the synthesis of HIV-1 protease inhibitors. It is also used in the synthesis of oxazoles, cyclohexanone derivatives as analgesic and antimicrobial agents.
Solubility
Soluble in methanol. Reacts with water forming Hcl.
Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases. Store at 2 - 8°C.
Specifications
Melting Point | 38°C to 42°C |
Boiling Point | 246°C |
Flash Point | >110°C (230°F) |
Quantity | 50 g |
UN Number | UN3261 |
Beilstein | 636641 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in methanol. Reacts with water forming Hcl. |
Formula Weight | 219.47 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'