missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzoic hydrazide 98%, ACROS Organics™

Brand:  Acros Organics 386460010

Code : 29

Additional Details : CAS Number : 5933-32-4 Weight : 0.05100kg


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10075503

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

4-Bromobenzoic hydrazide
97.5
98%
C7H7BrN2O
215.05
4-Bromobenzhydrazide
5933-32-4
100.0
98%
BrC6H4C(O)NHNH2
MFCD00007602

Specifications

4-Bromobenzoic hydrazide
Authentic
100.0
Light beige
1g
98%
C7H7BrN2O
215.05
4-Bromobenzhydrazide
5933-32-4
97.5
Glass bottle
166.0° to 168.0°C
Powder
98%
BrC6H4C(O)NHNH2
MFCD00007602
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Bromobenzoic hydrazide

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS