missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7020530
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
586-76-5
C7H5BrO2
201.02
MFCD00002529
TUXYZHVUPGXXQG-UHFFFAOYSA-N
p-bromobenzoic acid, benzoic acid, 4-bromo, p-carboxybromobenzene, benzoic acid, p-bromo, p-bromobenzenecarboxylic acid, 4-bromo-benzoic acid, 4-brom-benzoesaeure, 4-carboxybromobenzene, para bromo benzoic acid, 4bromo-benzoic acid
11464
CHEBI:60698
4-bromobenzoic acid
OC(=O)C1=CC=C(Br)C=C1
This item is not returnable. View return policy
586-76-5
C7H5BrO2
201.02
MFCD00002529
TUXYZHVUPGXXQG-UHFFFAOYSA-N
p-bromobenzoic acid, benzoic acid, 4-bromo, p-carboxybromobenzene, benzoic acid, p-bromo, p-bromobenzenecarboxylic acid, 4-bromo-benzoic acid, 4-brom-benzoesaeure, 4-carboxybromobenzene, para bromo benzoic acid, 4bromo-benzoic acid
11464
CHEBI:60698
4-bromobenzoic acid
OC(=O)C1=CC=C(Br)C=C1

4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.

Solubility
Slightly soluble in hot water. Soluble in alcohol and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 253°C to 258°C
Density 1.894
Odor Odorless
Quantity 25 g
Beilstein 1906923
Merck Index 14,1408
Solubility Information Slightly soluble in hot water. Soluble in alcohol and ether.
Formula Weight 201.02
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'