missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromobenzo-18-crown-6, 98%, ACROS Organics™

Brand:  Acros Organics 357230050

Code : 29

Additional Details : CAS Number : 75460-28-5 Weight : 0.05500kg

Quantity 5g

 View more versions of this product

Product Code. 10764743

  • £134.70 / 5g


Estimated Shipment
27-02-2018
Add to basket

Specifications

Chemical Identifiers

4'-Bromobenzo-18-crown-6
97.5
98%
C16H23BrO6
MFCD00143200
75460-28-5
100.0
98%
391.26

Specifications

4'-Bromobenzo-18-crown-6
Authentic
100.0
White
5g
98%
C16H23BrO6
MFCD00143200
75460-28-5
97.5
Glass bottle
79.0° to 83.0°C
Powder
98%
391.26
Videos
SDS