missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde diethyl acetal, 99%, Thermo Scientific™

Catalog Number p-3598203
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
34421-94-8
C11H15BrO2
259.14
MFCD01863514
BFSNEBVTOODGHZ-UHFFFAOYSA-N
4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal
688340
1-bromo-4-(diethoxymethyl)benzene
CCOC(OCC)C1=CC=C(Br)C=C1
This item is not returnable. View return policy
34421-94-8
C11H15BrO2
259.14
MFCD01863514
BFSNEBVTOODGHZ-UHFFFAOYSA-N
4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal
688340
1-bromo-4-(diethoxymethyl)benzene
CCOC(OCC)C1=CC=C(Br)C=C1
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2740g/mL
Boiling Point 75°C (0.2 mmHg)
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH(OCH2CH3)2
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Quantity 10 g
Specific Gravity 1.274
Formula Weight 259.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromobenzaldehyde diethyl acetal
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobenzaldehyde diethyl acetal
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'