missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog Number p-4229098
38.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
1122-91-4
C7H5BrO
185.02
ZRYZBQLXDKPBDU-UHFFFAOYSA-N
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
70741
4-bromobenzaldehyde
C1=CC(=CC=C1C=O)Br
1122-91-4
C7H5BrO
185.02
ZRYZBQLXDKPBDU-UHFFFAOYSA-N
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
70741
4-bromobenzaldehyde
C1=CC(=CC=C1C=O)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 55.0°C to 58.0°C
Color White
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CHO
Quantity 25 g
Beilstein 07,239
Solubility Information Solubility in water: insoluble
Formula Weight 185.02
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'