missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoanisole, 99%, Thermo Scientific Chemicals

Catalog Number p-7081729
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
104-92-7
C7H7BrO
187.036
MFCD00000097
QJPJQTDYNZXKQF-UHFFFAOYSA-N
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
7730
CHEBI:47257
1-bromo-4-methoxybenzene
COC1=CC=C(C=C1)Br
This item is not returnable. View return policy
104-92-7
C7H7BrO
187.036
MFCD00000097
QJPJQTDYNZXKQF-UHFFFAOYSA-N
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
7730
CHEBI:47257
1-bromo-4-methoxybenzene
COC1=CC=C(C=C1)Br

4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point 10°C to 13°C
Density 1.494
Boiling Point 98°C to 99°C (10 mm)
Flash Point 94°C (201°F)
Assay Percent Range 99%
Refractive Index 1.564
Quantity 100 g
Beilstein 1237590
Merck Index 14,1427
Solubility Information Immiscible with water.
Formula Weight 187.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromoanisole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'