missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoaniline, 96%, pract., Thermo Scientific Chemicals

Catalog Number 10710482
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy
106-40-1
C6H6BrN
172.02
MFCD00007822
WDFQBORIUYODSI-UHFFFAOYSA-N
p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
7807
4-bromoaniline
C1=CC(=CC=C1N)Br
This item is not returnable. View return policy
106-40-1
C6H6BrN
172.02
MFCD00007822
WDFQBORIUYODSI-UHFFFAOYSA-N
p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
7807
4-bromoaniline
C1=CC(=CC=C1N)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 60.0°C to 64.0°C
Color Brown to Gray
Density 1.4970g/mL
Boiling Point 230.0°C to 250.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4NH2
Quantity 500 g
Beilstein 12,636
Merck Index 15,1411
Specific Gravity 1.497
Solubility Information Solubility in water: < 1 g / l. Other solubilities: soluble in most organic solvents except cold,petroleum,very soluble in alcohol and ether
Formula Weight 172.02
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromoaniline, (pract), 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'