missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoaniline, 96%, pract., ACROS Organics™

Brand:  Acros Organics 402870025

Code : 29

Additional Details : CAS Number : 106-40-1 Weight : 4.00000kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 2.5kg

 View more versions of this product

Product Code. 11995211

  • £282.60 / 2.50kg


Estimated Shipment
12-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromoaniline
106-40-1
100.0
96%
BrC6H4NH2
MFCD00007822
(pract), 96%
95.0
96%
C6H6BrN
172.02

Specifications

4-Bromoaniline
106-40-1
1.4970g/mL
Authentic
1.497
96%
C6H6BrN
172.02
60.0° to 64.0°C
230.0° to 250.0°C
>110°C
(pract), 96%
95.0
100.0
Glass bottle
Crystals or Crystalline Powder
96%
BrC6H4NH2
MFCD00007822
Grayish to brown
2.5kg
Videos
SDS