missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-tert-butyl-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65083.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429366

  • £74.73 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-tert-butyl-2-fluorobenzenesulfonamide
310.18
MFCD27824720
23709742

Specifications

4-Bromo-N-tert-butyl-2-fluorobenzenesulfonamide
1g
310.18
23709742
Solid
97%
MFCD27824720
Videos
SDS