missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-phenyl-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63710.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15408466

  • £398.56 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-phenyl-3-(trifluoromethyl)benzenesulfonamide
380.18
MFCD26143383

Specifications

4-Bromo-N-phenyl-3-(trifluoromethyl)benzenesulfonamide
5g
380.18
Solid
97%
MFCD26143383
Videos
SDS