missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N,N-bis(4-methoxybenzyl)benzenesulfonamide, 95%, Thermo Scientific™

Catalog Number 15449046
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
192767-23-0
C22H22BrNO4S
476.385
MFCD27665298
YYLDNQJUMNGLLQ-UHFFFAOYSA-N
n,n-bis 4-methoxybenzyl-4-bromobenzenesulfonamide, 4-bromo-n,n-bis 4-methoxybenzyl benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzene-1-sulfonamide, benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzenesulfonamide
25138881
4-bromo-N,N-bis[(4-methoxyphenyl)methyl]benzenesulfonamide
COC1=CC=C(C=C1)CN(CC2=CC=C(C=C2)OC)S(=O)(=O)C3=CC=C(C=C3)Br
This item is not returnable. View return policy
192767-23-0
C22H22BrNO4S
476.385
MFCD27665298
YYLDNQJUMNGLLQ-UHFFFAOYSA-N
n,n-bis 4-methoxybenzyl-4-bromobenzenesulfonamide, 4-bromo-n,n-bis 4-methoxybenzyl benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzene-1-sulfonamide, benzenesulfonamide, 4-bromo-n,n-bis 4-methoxyphenyl methyl, 4-bromo-n,n-bis 4-methoxyphenyl methyl benzenesulfonamide
25138881
4-bromo-N,N-bis[(4-methoxyphenyl)methyl]benzenesulfonamide
COC1=CC=C(C=C1)CN(CC2=CC=C(C=C2)OC)S(=O)(=O)C3=CC=C(C=C3)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 476.38
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Bromo-N,N-bis(4-methoxybenzyl)benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'