Learn More
Alfa Aesar™ 4-Bromo-N-cyclopropyl-2-fluorobenzenesulfonamide, 97%
Brand: Alfa Aesar™ H64987.03
Code : NEW
Additional Details : CAS Number : 1055995-89-5 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4-Bromo-N-cyclopropyl-2-fluorobenzenesulfonamide | |
1055995-89-5 | |
294.14 | |
MFCD27975706 |
Specifications
4-Bromo-N-cyclopropyl-2-fluorobenzenesulfonamide | |
Solid | |
97% | |
MFCD27975706 |
1055995-89-5 | |
1g | |
294.14 |