missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-cyclopentyl-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64549.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15428976

  • £91.90 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-cyclopentyl-2-fluorobenzenesulfonamide
322.19
MFCD21364216

Specifications

4-Bromo-N-cyclopentyl-2-fluorobenzenesulfonamide
1g
322.19
Solid
97%
MFCD21364216
Videos
SDS