missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-cyclohexyl-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62143.06

Code : NEW

Additional Details : CAS Number : 1020253-02-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15497076

  • £398.56 / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-cyclohexyl-3-(trifluoromethyl)benzenesulfonamide
1020253-02-4
386.23
MFCD09972158

Specifications

4-Bromo-N-cyclohexyl-3-(trifluoromethyl)benzenesulfonamide
Solid
97%
MFCD09972158
1020253-02-4
5g
386.23
Videos
SDS