missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-cyclohexyl-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64886.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419236

  • £99.64 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-cyclohexyl-2-fluorobenzenesulfonamide
336.22
MFCD27392032

Specifications

4-Bromo-N-cyclohexyl-2-fluorobenzenesulfonamide
1g
336.22
Solid
97%
MFCD27392032
Videos
SDS