missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(5-chloro-2-methylphenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64871.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459216

  • £46.00 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(5-chloro-2-methylphenyl)-2-fluorobenzenesulfonamide
378.64
MFCD28098732

Specifications

4-Bromo-N-(5-chloro-2-methylphenyl)-2-fluorobenzenesulfonamide
1g
378.64
Solid
97%
MFCD28098732
Videos
SDS