missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(5-chloro-2-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63828.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405645

  • £47.81 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(5-chloro-2-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
444.65
MFCD26143387

Specifications

4-Bromo-N-(5-chloro-2-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
444.65
Solid
97%
MFCD26143387
Videos
SDS