missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-n-butylphenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65815.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459926

  • £91.90 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-n-butylphenyl)-2-fluorobenzenesulfonamide
386.28
MFCD27976899

Specifications

4-Bromo-N-(4-n-butylphenyl)-2-fluorobenzenesulfonamide
1g
386.28
Solid
97%
MFCD27976899
Videos
SDS