Alfa Aesar™ 4-Bromo-N-(4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H63654.MD

250MG 4-Bromo-N-(4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97% 250mg

UNSPSC: 12352105

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15468426

Quantity Price
1 £ 23.96 / 250mg
Estimated Shipment date
from Supplier 07-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Bromo-N-(4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
Formula Weight 410.21
MDL Number MFCD26143379

Specification

Formula Weight 410.21
Chemical Name or Material 4-Bromo-N-(4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
Physical Form Solid
Quantity 250mg
Assay 97%
MDL Number MFCD26143379