missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-methoxybenzyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63015.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15428096

  • £59.78 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-methoxybenzyl)-3-(trifluoromethyl)benzenesulfonamide
424.23
MFCD26143352

Specifications

4-Bromo-N-(4-methoxybenzyl)-3-(trifluoromethyl)benzenesulfonamide
1g
424.23
Solid
97%
MFCD26143352
Videos
SDS