missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62184.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447136

  • £68.26 / 1g


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
398.17
MFCD26143313

Specifications

4-Bromo-N-(4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
97%
398.17
Solid
1g
MFCD26143313
Videos
SDS