missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-fluorobenzyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62462.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417466

  • £73.56 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-fluorobenzyl)-3-(trifluoromethyl)benzenesulfonamide
412.2
MFCD26143326

Specifications

4-Bromo-N-(4-fluorobenzyl)-3-(trifluoromethyl)benzenesulfonamide
97%
412.2
Solid
1g
MFCD26143326
Videos
SDS