missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-fluorobenzyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62462.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15427466

  • £367.82 / 5g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-fluorobenzyl)-3-(trifluoromethyl)benzenesulfonamide
412.2
MFCD26143326

Specifications

4-Bromo-N-(4-fluorobenzyl)-3-(trifluoromethyl)benzenesulfonamide
97%
412.2
Solid
5g
MFCD26143326
Videos
SDS