missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-[4-chloro-3-(trifluoromethyl)phenyl]-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65639.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429796

  • £79.71 / 1g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-[4-chloro-3-(trifluoromethyl)phenyl]-2-fluorobenzenesulfonamide
432.61
MFCD27975794

Specifications

4-Bromo-N-[4-chloro-3-(trifluoromethyl)phenyl]-2-fluorobenzenesulfonamide
1g
432.61
Solid
97%
MFCD27975794
Videos
SDS