missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-chloro-2-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62224.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447176

  • £56.29 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-chloro-2-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
428.65
MFCD26143315

Specifications

4-Bromo-N-(4-chloro-2-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
428.65
Solid
97%
MFCD26143315
Videos
SDS