missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-bromobenzyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64388.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498876

  • £99.64 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-bromobenzyl)-2-fluorobenzenesulfonamide
423.1
MFCD28098724

Specifications

4-Bromo-N-(4-bromobenzyl)-2-fluorobenzenesulfonamide
1g
423.1
Solid
97%
MFCD28098724
Videos
SDS