missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-bromo-3-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62827.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447886

  • £59.78 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-bromo-3-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
473.1
MFCD26143345

Specifications

4-Bromo-N-(4-bromo-3-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
473.1
Solid
97%
MFCD26143345
Videos
SDS