missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-bromo-3-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62827.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15457886

  • £298.92 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-bromo-3-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
473.1
MFCD26143345

Specifications

4-Bromo-N-(4-bromo-3-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
473.1
Solid
97%
MFCD26143345
Videos
SDS