missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-bromo-2-methylphenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64796.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499146

  • £59.78 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-bromo-2-methylphenyl)-2-fluorobenzenesulfonamide
423.1
MFCD27975699

Specifications

4-Bromo-N-(4-bromo-2-methylphenyl)-2-fluorobenzenesulfonamide
1g
423.1
Solid
97%
MFCD27975699
Videos
SDS