missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-[3-(trifluoromethyl)phenyl]benzamide, 97%, Thermo Scientific™

Catalog Number 15413936
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
56661-32-6
C14H9BrF3NO
344.131
MFCD00421306
URMINAWTHPPDQD-UHFFFAOYSA-N
4-bromo-n-3-trifluoromethyl phenyl benzamide, benzamide, 4-bromo-n-3-trifluoromethyl phenyl, 4-bromophenyl-n-3-trifluoromethyl phenyl carboxamide
788568
4-bromo-N-[3-(trifluoromethyl)phenyl]benzamide
C1=CC(=CC(=C1)NC(=O)C2=CC=C(C=C2)Br)C(F)(F)F
This item is not returnable. View return policy
56661-32-6
C14H9BrF3NO
344.131
MFCD00421306
URMINAWTHPPDQD-UHFFFAOYSA-N
4-bromo-n-3-trifluoromethyl phenyl benzamide, benzamide, 4-bromo-n-3-trifluoromethyl phenyl, 4-bromophenyl-n-3-trifluoromethyl phenyl carboxamide
788568
4-bromo-N-[3-(trifluoromethyl)phenyl]benzamide
C1=CC(=CC(=C1)NC(=O)C2=CC=C(C=C2)Br)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in a dry, cool conditions in a well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 100 mg
Formula Weight 344.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Bromo-N-[3-(trifluoromethyl)phenyl]benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'