missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(3-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62711.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15497756

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

4-Bromo-N-(3-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
394.21
MFCD26143335

Specifications

4-Bromo-N-(3-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
394.21
Solid
97%
MFCD26143335
Videos
SDS