missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(3-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63497.03

UNSPSC : 12352105

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468336

  • £95.61 / 1g


Estimated Shipment date
from Supplier 09-10-2017
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(3-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
410.21
MFCD26143370

Specifications

4-Bromo-N-(3-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
410.21
Solid
97%
MFCD26143370
Documentation

Technical Documents