Alfa Aesar™ 4-Bromo-N-(3-fluorobenzyl)-3-(trifluoromethyl)benzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H62770.MD

UNSPSC: 12352105

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15477826

Quantity Price
1 £ 21.2 / 250mg
Estimated Shipment date
from Supplier 11-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Bromo-N-(3-fluorobenzyl)-3-(trifluoromethyl)benzenesulfonamide
Formula Weight 412.2
MDL Number MFCD26143339

Specification

Formula Weight 412.2
Chemical Name or Material 4-Bromo-N-(3-fluorobenzyl)-3-(trifluoromethyl)benzenesulfonamide
Physical Form Solid
MDL Number MFCD26143339
Assay 97%
Quantity 250mg
Documentation

Technical documents