missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Bromo-N-(3-chloro-4-methylphenyl)-5-fluorobenzenesulfonamide, 97%
Brand: Alfa Aesar™ H61662.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4-Bromo-N-(3-chloro-4-methylphenyl)-5-fluorobenzenesulfonamide | |
C13H10BrClFNO2S | |
378.64 | |
MFCD24848996 |
Specifications
4-Bromo-N-(3-chloro-4-methylphenyl)-5-fluorobenzenesulfonamide | |
C13H10BrClFNO2S | |
MFCD24848996 |
97% | |
378.64 | |
1g |